IITZ-01 97%

Supplier: Ambeed
Warning

Synonyms: IITZ01, N2-(4-(1H-benzo[d]imidazol-2-yl)phenyl)-N4-(4-fluorophenyl)-6-morpholino-1,3,5-triazine-2,4-diamine

A888602-10MG A888602-1MG A888602-25MG A888602-50MG A888602-5MG
76654-956EA 41.38 USD
76654-956 76654-958 76654-960 76654-962 76654-964
IITZ-01 97%
IITZ-01

Precaución: for Research Use Only

Formula: C₂₆H₂₃FN₈O
MW: 482.51 g/mol
MDL Number: MFCD31746904
CAS Number: 1807988-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR