rel-(1r,4r)-4-((2-Phenyl-7H-pyrrolo[2,3-d]pyrimidin-4-yl)amino)cyclohexan-1-ol 98%

Supplier: Ambeed
Warning

Synonyms: Derenofylline

A887654-10MG A887654-1MG A887654-5MG
76654-940EA 28.65 USD
76654-940 76654-942 76654-944
rel-(1r,4r)-4-((2-Phenyl-7H-pyrrolo[2,3-d]pyrimidin-4-yl)amino)cyclohexan-1-ol 98%
rel-(1r,4r)-4-((2-Phenyl-7H-pyrrolo[2,3-d]pyrimidin-4-yl)amino)cyclohexan-1-ol

Precaución: for Research Use Only

Formula: C₁₈H₂₀N₄O
MW: 308.3776 g/mol
MDL Number: MFCD12756289
CAS Number: 251945-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR