Autophinib 97%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-N-(5-methyl-1H-pyrazol-3-yl)-2-(4-nitrophenoxy)-pyrimidinamine, 6-Chloro-N-(5-methyl-1H-pyrazol-3-yl)-2-(4-nitrophenoxy)pyrimidin-4-amine

A886466-10MG A886466-1MG A886466-2MG A886466-50MG A886466-5MG
76654-916EA 22.28 USD
76654-916 76654-918 76654-920 76654-922 76654-924
Autophinib 97%
Autophinib

Precaución: for Research Use Only

Formula: C₁₄H₁₁ClN₆O₃
MW: 346.73 g/mol
MDL Number: MFCD31560472
CAS Number: 1644443-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR