Iron(III) stearate 98%

Supplier: Ambeed
Warning

Synonyms: Ferric Stearate, Iron(III) Stearate, Stearic Acid Iron(III) Salt

A886464-100G A886464-25G A886464-500G A886464-5G
76654-908EA 50 USD
76654-908 76654-910 76654-912 76654-914
Iron(III) stearate 98%
Iron(III) stearate

Precaución: for Research Use Only

Formula: C₁₈H₃₆O₂
MW: 906.27 g/mol
MDL Number: MFCD00050716
CAS Number: 555-36-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR