4,5-Dichloro-2-ethyl-2,3-dihydropyridazin-3-one 95%

Supplier: Ambeed
Danger

A885889-100MG A885889-250MG
76654-888EA 112.46 USD
76654-888 76654-890
4,5-Dichloro-2-ethyl-2,3-dihydropyridazin-3-one 95%
4,5-Dichloro-2-ethyl-2,3-dihydropyridazin-3-one

Precaución: for Research Use Only

Formula: C₆H₆Cl₂N₂O
MW: 193.031 g/mol
MDL Number: MFCD00793799
CAS Number: 33098-10-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR