1,12-Diacetoxydodecane 98% (by GC)

Supplier: Ambeed
Warning

Synonyms: Dodecamethylene Diacetate, 1,12-Dodecanediol Diacetate

A885538-1G A885538-250MG A885538-5G
76654-878EA 19.1 USD
76654-878 76654-880 76654-882
1,12-Diacetoxydodecane 98% (by GC)
1,12-Diacetoxydodecane

Precaución: for Research Use Only

Formula: C₁₆H₃₀O₄
MW: 286.41 g/mol
MDL Number: MFCD06252255
CAS Number: 42236-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR