Isopropyl methoxyacetate 98%

Supplier: Ambeed
Warning

A885230-25G A885230-5G
76654-866EA 26.53 USD
76654-866 77749-691
Isopropyl methoxyacetate 98%
Isopropyl methoxyacetate

Precaución: for Research Use Only

Formula: C₆H₁₂O₃
MW: 132.1577 g/mol
MDL Number: MFCD16038497
CAS Number: 17640-21-0
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR