5,6,11,12-Tetradehydrodibenzo[a,e]cyclooctene 95%

Supplier: Ambeed
Warning

Synonyms: Dibenzo[a,e]cyclooctadien-5,11-diyne, 5,6,11,12-Tetradehydrodibenzo[a,e]cyclooctatetraene, 5,6,11,12-Tetradehydrodibenzo[a,e][8]annulene

A882663-100MG A882663-250MG A882663-50MG A882663-1G
76654-842EA 169 USD
76654-842 77618-472 77618-474 77749-685
5,6,11,12-Tetradehydrodibenzo[a,e]cyclooctene 95%
5,6,11,12-Tetradehydrodibenzo[a,e]cyclooctene

Precaución: for Research Use Only

Formula: C₁₆H₈
MDL Number: MFCD28386121
CAS Number: 53397-65-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR