1-Benzyl-1H-indazol-3-amine 97%

Supplier: Ambeed
Warning

A879077-100MG A879077-250MG
76654-822EA 60 USD
76654-822 76654-824
1-Benzyl-1H-indazol-3-amine 97%
1-Benzyl-1H-indazol-3-amine

Precaución: for Research Use Only

Formula: C₁₄H₁₃N₃
MW: 223.2700 g/mol
CAS Number: 28519-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR