3,6,9,12,15-Pentaoxaheptadecane-1,17-diyl diacrylate 98%

Supplier: Ambeed
Warning

Synonyms: Bis-acrylate-PEG6

A1350027-100MG A1350027-1G A1350027-250MG
76658-166EA 49.87 USD
76658-166 76658-168 76658-170
3,6,9,12,15-Pentaoxaheptadecane-1,17-diyl diacrylate 98%
3,6,9,12,15-Pentaoxaheptadecane-1,17-diyl diacrylate

Precaución: for Research Use Only

Formula: C₁₈H₃₀O₉
MW: 390.4254 g/mol
MDL Number: MFCD32706451
CAS Number: 85136-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR