1,11-Bis(maleimido)-3,6,9-trioxaundecane 95% (by HPLC)

Supplier: Ambeed
Warning

Synonyms: 1,11-Bis(maleimido)triethylene Glycol, BM-PEG3

A878286-1G A878286-250MG
76654-814EA 187 USD
76654-814 77623-096
1,11-Bis(maleimido)-3,6,9-trioxaundecane 95% (by HPLC)
1,11-Bis(maleimido)-3,6,9-trioxaundecane

Precaución: for Research Use Only

Formula: C₁₆H₂₀N₂O₇
MW: 352.34 g/mol
MDL Number: MFCD01863352
CAS Number: 86099-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR