Propylene glycol monostearate 83% (by GC)

Supplier: Ambeed
Warning

Synonyms: Hydroxypropyl Stearate, Stearic Acid Hydroxypropyl Ester

A877422-1G A877422-5G
76654-808EA 5.31 USD
76654-808 76654-810
Propylene glycol monostearate 83% (by GC)
Propylene glycol monostearate

Precaución: for Research Use Only

Formula: C₂₁H₄₂O₃
MW: 342.56 g/mol
MDL Number: MFCD00021902
CAS Number: 1323-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR