5-(Trifluoromethyl)-1-(2-fluorophenyl)-1H-pyrazole-4-carboxylic acid 97%

Supplier: Ambeed
Warning

A874250-100MG A874250-1G A874250-250MG
76654-764EA 47.75 USD
76654-764 76654-766 76654-768
5-(Trifluoromethyl)-1-(2-fluorophenyl)-1H-pyrazole-4-carboxylic acid 97%
5-(Trifluoromethyl)-1-(2-fluorophenyl)-1H-pyrazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₆F₄N₂O₂
MW: 274.1712 g/mol
MDL Number: MFCD08446378
CAS Number: 948293-85-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR