Methyl 2-hydroxy-2-phenylpropanoate 95%

Supplier: Ambeed
Warning

A874137-100MG A874137-250MG
76654-760EA 110.34 USD
76654-760 76654-762
Methyl 2-hydroxy-2-phenylpropanoate 95%
Methyl 2-hydroxy-2-phenylpropanoate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MW: 180.2005 g/mol
MDL Number: MFCD02683214
CAS Number: 20731-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR