TM-5275 sodium salt 98%

Supplier: Ambeed
Danger

Synonyms: 5-Chloro-2-[[2-[2-[4-(Diphenylmethyl)-1-piperazinyl]-2-oxoethoxy]acetyl]amino]benzoic acid sodium salt, Sodium 2-(2-(2-(4-benzhydrylpiperazin-1-yl)-2-oxoethoxy)acetamido)-5-chlorobenzoate

A873934-100MG A873934-10MG A873934-1MG A873934-25MG A873934-2MG A873934-50MG A873934-5MG
76654-746EA 608.96 USD
76654-746 76654-748 76654-750 76654-752 76654-754 76654-756 76654-758
TM-5275 sodium salt 98%
TM-5275 sodium salt

Precaución: for Research Use Only

Formula: C₂₈H₂₇ClN₃NaO₅
MW: 543.98 g/mol
MDL Number: MFCD28396414
CAS Number: 1103926-82-4
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR