2-Amino-5-fluoro-3-methylbenzoic acid 95%

Supplier: Ambeed
Warning

Synonyms: 5-Fluoro-3-methylanthranilic acid

A872205-100MG A872205-1G A872205-250MG A872205-5G
76654-732EA 30.77 USD
76654-732 76654-734 76654-736 77623-088
2-Amino-5-fluoro-3-methylbenzoic acid 95%
2-Amino-5-fluoro-3-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₈FNO₂
MW: 169.153 g/mol
MDL Number: MFCD06409107
CAS Number: 874804-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR