1-(2,5-Difluorophenyl)ethan-1-ol 95%

Supplier: Ambeed
Warning

A871517-1G A871517-250MG A871517-5G
76654-724EA 106 USD
76654-724 76654-726 76654-728
1-(2,5-Difluorophenyl)ethan-1-ol 95%
1-(2,5-Difluorophenyl)ethan-1-ol

Precaución: for Research Use Only

Formula: C₈H₈F₂O
MDL Number: MFCD08442989
CAS Number: 3874-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR