3-Chloro-p-toluidine hydrochloride 95%

Supplier: Ambeed
Warning

A871285-25G A871285-1G A871285-100G A871285-5G
76654-722EA 77 USD
76654-722 77623-086 77784-281 77784-282
3-Chloro-p-toluidine hydrochloride 95%
3-Chloro-p-toluidine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₉Cl₂N
MDL Number: MFCD00242925
CAS Number: 7745-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR