3-Cyclohexyl-DL-alanine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-3-cyclohexylpropanoic acid

A871133-1G A871133-250MG A871133-5G
76654-716EA 23.34 USD
76654-716 76654-718 76654-720
3-Cyclohexyl-DL-alanine 97%
3-Cyclohexyl-DL-alanine

Precaución: for Research Use Only

Formula: C₉H₁₇NO₂
MW: 171.237 g/mol
MDL Number: MFCD00996128
CAS Number: 4441-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR