tert-Butyl (3S,4R)-3-amino-4-phenylpyrrolidine-1-carboxylate 97%

Supplier: Ambeed
Warning

A870474-100MG A870474-1G A870474-250MG
76654-708EA 139 USD
76654-708 76654-710 76654-712
tert-Butyl (3S,4R)-3-amino-4-phenylpyrrolidine-1-carboxylate 97%
tert-Butyl (3S,4R)-3-amino-4-phenylpyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₅H₂₂N₂O₂
MDL Number: MFCD09755983
CAS Number: 1643979-48-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR