9-Benzoyl-3,6-dibromo-9H-carbazole 98% (by GC)

Supplier: Ambeed
Warning

A869979-1G A869979-250MG A869979-5G
76654-702EA 25.47 USD
76654-702 76654-704 76654-706
9-Benzoyl-3,6-dibromo-9H-carbazole 98% (by GC)
9-Benzoyl-3,6-dibromo-9H-carbazole

Precaución: for Research Use Only

Formula: C₁₉H₁₁Br₂NO
MW: 429.11 g/mol
MDL Number: MFCD30749253
CAS Number: 912850-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR