2-(2-Methylphenyl)piperazine dihydrochloride 95%

Supplier: Ambeed
Warning

A868958-1G A868958-250MG
76654-686EA 63 USD
76654-686 76654-688
2-(2-Methylphenyl)piperazine dihydrochloride 95%
2-(2-Methylphenyl)piperazine dihydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₈Cl₂N₂
MW: 249.1800 g/mol
MDL Number: MFCD30539794
CAS Number: 161115-88-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR