2-(2,4-Difluorophenyl)-2,2-difluoroethan-1-amine hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A867740-100MG
A867740-1G
A867740-250MG
A867740-50MG
76654-672EA
103.97
USD
76654-672
76654-674
76654-676
76654-678
2-(2,4-Difluorophenyl)-2,2-difluoroethan-1-amine hydrochloride 95%
2-(2,4-Difluorophenyl)-2,2-difluoroethan-1-amine hydrochloride
Precaución: for Research Use Only
Formula:
C₈H₈ClF₄N MW: 229.6024 g/mol |
MDL Number:
MFCD30502238 CAS Number: 2060062-92-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...