3-(Difluoromethyl)thiophene-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A867528-100MG A867528-1G A867528-250MG
76654-666EA 74.27 USD
76654-666 76654-668 76654-670
3-(Difluoromethyl)thiophene-2-carboxylic acid 98%
3-(Difluoromethyl)thiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₄F₂O₂S
MW: 178.1566 g/mol
MDL Number: MFCD20659264
CAS Number: 189330-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR