1,1,1,3,3,3-Hexafluoropropan-2-yl 4-(2-(pyrrolidin-1-yl)-4-(trifluoromethyl)benzyl)piperazine-1-carboxylate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Elcubragistat
A866363-10MG
A866363-1MG
A866363-5MG
76654-646EA
48.81
USD
76654-646
76654-648
76654-650
1,1,1,3,3,3-Hexafluoropropan-2-yl 4-(2-(pyrrolidin-1-yl)-4-(trifluoromethyl)benzyl)piperazine-1-carboxylate 98%
1,1,1,3,3,3-Hexafluoropropan-2-yl 4-(2-(pyrrolidin-1-yl)-4-(trifluoromethyl)benzyl)piperazine-1-carboxylate
Precaución: for Research Use Only
Formula:
C₂₀H₂₂F₉N₃O₂ MW: 507.39 g/mol |
MDL Number:
MFCD31735112 CAS Number: 1446817-84-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...