1-Propylpiperidin-4-amine dihydrochloride 95%

Supplier: Ambeed
Warning

A866175-100MG A866175-1G A866175-250MG
76654-640EA 14.86 USD
76654-640 76654-642 76654-644
1-Propylpiperidin-4-amine dihydrochloride 95%
1-Propylpiperidin-4-amine dihydrochloride

Precaución: for Research Use Only

Formula: C₈H₂₀Cl₂N₂
MW: 215.1638 g/mol
MDL Number: MFCD09759213
CAS Number: 1158561-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR