3-Amino-2-bromo-6-fluorobenzonitrile 97%

Supplier: Ambeed
Danger

A865619-100MG A865619-250MG
76654-624EA 133.68 USD
76654-624 76654-626
3-Amino-2-bromo-6-fluorobenzonitrile 97%
3-Amino-2-bromo-6-fluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄BrFN₂
MW: 215.0225 g/mol
MDL Number: MFCD30469059
CAS Number: 1415633-90-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR