4-Formyl-3,5-dimethyl-1H-pyrrole-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A865491-100MG A865491-250MG
76654-620EA 114 USD
76654-620 76654-622
4-Formyl-3,5-dimethyl-1H-pyrrole-2-carboxylic acid 95%
4-Formyl-3,5-dimethyl-1H-pyrrole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.1620 g/mol
MDL Number: MFCD00030406
CAS Number: 17120-26-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR