Ethyl 6-bromoimidazo[1,2-a]pyrimidine-2-carboxylate 95%

Supplier: Ambeed
Warning

A865463-1G A865463-250MG A865463-100MG
76654-616EA 192.03 USD
76654-616 76654-618 77623-068
Ethyl 6-bromoimidazo[1,2-a]pyrimidine-2-carboxylate 95%
Ethyl 6-bromoimidazo[1,2-a]pyrimidine-2-carboxylate

Precaución: for Research Use Only

Formula: C₉H₈BrN₃O₂
MW: 270.0827 g/mol
MDL Number: MFCD09838962
CAS Number: 944896-67-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR