2-Methyl-4-(morpholin-4-yl)butan-2-amine dihydrochloride 95%

Supplier: Ambeed
Warning

A865328-100MG A865328-250MG
76654-606EA 89 USD
76654-606 76654-608
2-Methyl-4-(morpholin-4-yl)butan-2-amine dihydrochloride 95%
2-Methyl-4-(morpholin-4-yl)butan-2-amine dihydrochloride

Precaución: for Research Use Only

Formula: C₉H₂₂Cl₂N₂O
MW: 245.1898 g/mol
MDL Number: MFCD30476234
CAS Number: 2044927-06-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR