tert-Butyl (7S)-7-amino-5-azaspiro[2.4]heptane-5-carboxylate 97%

Supplier: Ambeed
Danger

A864860-1G A864860-250MG
76654-600EA 463 USD
76654-600 76654-602
tert-Butyl (7S)-7-amino-5-azaspiro[2.4]heptane-5-carboxylate 97%
tert-Butyl (7S)-7-amino-5-azaspiro[2.4]heptane-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₀N₂O₂
MDL Number: MFCD28501843
CAS Number: 1638744-35-0
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR