3-(Piperazin-1-yl)-1,2,4-triazine 95%

Supplier: Ambeed
Warning

A864560-1G A864560-250MG
76654-594EA 395 USD
76654-594 76654-596
3-(Piperazin-1-yl)-1,2,4-triazine 95%
3-(Piperazin-1-yl)-1,2,4-triazine

Precaución: for Research Use Only

Formula: C₇H₁₁N₅
MDL Number: MFCD11108269
CAS Number: 341010-36-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR