7,8-Difluoroquinoline 98%

Supplier: Ambeed
Warning

A863756-1G A863756-250MG A863756-5G
76654-582EA 41 USD
76654-582 76654-584 76654-586
7,8-Difluoroquinoline 98%
7,8-Difluoroquinoline

Precaución: for Research Use Only

Formula: C₉H₅F₂N
MW: 165.1395 g/mol
MDL Number: MFCD01075229
CAS Number: 145241-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR