4-Bromobutan-2-ol 95%

Supplier: Ambeed
Danger

A863596-100MG A863596-250MG
76654-578EA 25 USD
76654-578 76654-580
4-Bromobutan-2-ol 95%
4-Bromobutan-2-ol

Precaución: for Research Use Only

Formula: C₄H₉BrO
MW: 153.0177 g/mol
MDL Number: MFCD20621201
CAS Number: 6089-17-4
UN: 1987
ADR: 3 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR