6-(Benzyloxy)-2-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 96%
Supplier: Ambeed
|
Warning
|
A863454-100MG
A863454-1G
A863454-250MG
A863454-5G
76654-570EA
36.08
USD
76654-570
76654-572
76654-574
76654-576
6-(Benzyloxy)-2-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 96%
6-(Benzyloxy)-2-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Precaución: for Research Use Only
Formula:
C₁₉H₂₄BNO₃ MW: 325.2098 g/mol |
MDL Number:
MFCD16996688 CAS Number: 2096337-41-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...