Methyl 5-tert-butyl-1,3,4-oxadiazole-2-carboxylate 95%

Supplier: Ambeed
Warning

A863301-100MG A863301-250MG
76654-566EA 91.24 USD
76654-566 76654-568
Methyl 5-tert-butyl-1,3,4-oxadiazole-2-carboxylate 95%
Methyl 5-tert-butyl-1,3,4-oxadiazole-2-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₂N₂O₃
MW: 184.1925 g/mol
MDL Number: MFCD26058698
CAS Number: 1565976-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR