Ethylhydrazine dihydrochloride 95%

Supplier: Ambeed
Warning

A862803-1G A862803-5G A862803-25G
76654-562EA 9 USD
76654-562 76654-564 77784-258
Ethylhydrazine dihydrochloride 95%
Ethylhydrazine dihydrochloride

Precaución: for Research Use Only

Formula: C₂H₁₀Cl₂N₂
MDL Number: MFCD28118878
CAS Number: 49540-34-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR