5-Bromo-6-fluoroindoline 97%

Supplier: Ambeed
Warning

A862691-100MG A862691-250MG A862691-1G
76654-558EA 63 USD
76654-558 76654-560 77618-432
5-Bromo-6-fluoroindoline 97%
5-Bromo-6-fluoroindoline

Precaución: for Research Use Only

Formula: C₈H₇BrFN
MDL Number: MFCD22037765
CAS Number: 1368323-85-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR