INCB057643 99%

Supplier: Ambeed
Warning

Synonyms: 2,2,4-trimethyl-8-(6-methyl-7-oxo-6,7-dihydro-1H-pyrrolo[2,3-c]pyridin-4-yl)-6-(methylsulphonyl)-2H-benzo[b][1,4]oxazin-3(4H)-one, INCB-057643

A861913-100MG A861913-10MG A861913-1MG A861913-25MG A861913-50MG A861913-5MG
76654-538EA 411.63 USD
76654-538 76654-540 76654-542 76654-544 76654-546 76654-548
INCB057643 99%
INCB057643

Precaución: for Research Use Only

Formula: C₂₀H₂₁N₃O₅S
MW: 415.46 g/mol
MDL Number: MFCD31657400
CAS Number: 1820889-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR