(1R)-1-(Pyrazin-2-yl)ethan-1-amine dihydrochloride 95%

Supplier: Ambeed
Warning

A861037-100MG A861037-1G A861037-250MG
76654-526EA 67.9 USD
76654-526 76654-528 76654-530
(1R)-1-(Pyrazin-2-yl)ethan-1-amine dihydrochloride 95%
(1R)-1-(Pyrazin-2-yl)ethan-1-amine dihydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₁Cl₂N₃
MW: 196.0800 g/mol
MDL Number: MFCD29991525
CAS Number: 1955474-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR