NCGC00244536 95%

Supplier: Ambeed
Warning

Synonyms: 3-(8-hydroxyquinolin-6-yl)-N-(3-phenylpropyl)benzamide

A860757-10MG A860757-1MG A860757-5MG
76654-520EA 48.81 USD
76654-520 76654-522 76654-524
NCGC00244536 95%
NCGC00244536

Precaución: for Research Use Only

Formula: C₂₅H₂₂N₂O₂
MW: 382.46 g/mol
MDL Number: MFCD31382180
CAS Number: 2003260-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR