(3,4-Dihydro-2H-benzo[b][1,4]dioxepin-6-yl)boronic acid 95%

Supplier: Ambeed
Warning

A860625-1G A860625-250MG A860625-5G
76654-514EA 160.2 USD
76654-514 76654-516 76654-518
(3,4-Dihydro-2H-benzo[b][1,4]dioxepin-6-yl)boronic acid 95%
(3,4-Dihydro-2H-benzo[b][1,4]dioxepin-6-yl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BO₄
MW: 193.9922 g/mol
MDL Number: MFCD06801681
CAS Number: 1107064-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR