(R)-5-(2-(2,5-Difluorophenyl)pyrrolidin-1-yl)pyrazolo[1,5-a]pyrimidin-3-amine 97%

Supplier: Ambeed
Warning

A860346-1G A860346-250MG
76654-504EA 84.88 USD
76654-504 76654-506
(R)-5-(2-(2,5-Difluorophenyl)pyrrolidin-1-yl)pyrazolo[1,5-a]pyrimidin-3-amine 97%
(R)-5-(2-(2,5-Difluorophenyl)pyrrolidin-1-yl)pyrazolo[1,5-a]pyrimidin-3-amine

Precaución: for Research Use Only

Formula: C₁₆H₁₅F₂N₅
MW: 315.3206 g/mol
MDL Number: MFCD30802505
CAS Number: 1223404-88-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR