2H,3H-Naphtho[1,2-b]furan-2-one 95%

Supplier: Ambeed
Warning

A859261-100MG A859261-1G A859261-250MG
76654-490EA 196.27 USD
76654-490 76654-492 76654-494
2H,3H-Naphtho[1,2-b]furan-2-one 95%
2H,3H-Naphtho[1,2-b]furan-2-one

Precaución: for Research Use Only

Formula: C₁₂H₈O₂
MW: 184.1907 g/mol
MDL Number: MFCD29763014
CAS Number: 6050-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR