7-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[d]oxazole 95%

Supplier: Ambeed
Warning

A858388-1G A858388-250MG A858388-5G
76654-474EA 166.57 USD
76654-474 76654-476 76654-478
7-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[d]oxazole 95%
7-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[d]oxazole

Precaución: for Research Use Only

Formula: C₁₃H₁₆BNO₃
MW: 245.0820 g/mol
MDL Number: MFCD16995817
CAS Number: 2137501-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR