4-(2-Amino-4-ethyl-5-(1H-indazol-5-yl)pyridin-3-yl)phenol 98%

Supplier: Ambeed
Warning

Synonyms: GNE-6640

A858171-10MG A858171-1MG A858171-5MG
76654-468EA 114.58 USD
76654-468 76654-470 76654-472
4-(2-Amino-4-ethyl-5-(1H-indazol-5-yl)pyridin-3-yl)phenol 98%
4-(2-Amino-4-ethyl-5-(1H-indazol-5-yl)pyridin-3-yl)phenol

Precaución: for Research Use Only

Formula: C₂₀H₁₈N₄O
MW: 330.38 g/mol
MDL Number: MFCD31807618
CAS Number: 2009273-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR