2-(4-Hydroxyphenyl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione 95+%

Supplier: Ambeed
Warning

Synonyms: 4-Hydroxyphenylboronic acid MIDA ester

A857574-100G A857574-1G A857574-25G A857574-5G
76654-460EA 2005.11 USD
76654-460 76654-462 76654-464 76654-466
2-(4-Hydroxyphenyl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione 95+%
2-(4-Hydroxyphenyl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione

Precaución: for Research Use Only

Formula: C₁₁H₁₂BNO₅
MW: 249.028 g/mol
MDL Number: MFCD25541468
CAS Number: 1613586-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR