(S)-2-((4,6-Dimethoxypyrimidin-2-yl)oxy)-3-methoxy-3,3-diphenylpropanoic acid 99+%

Supplier: Ambeed
Warning

A857315-10MG A857315-1MG A857315-50MG A857315-5MG
76654-452EA 25.47 USD
76654-452 76654-454 76654-456 76654-458
(S)-2-((4,6-Dimethoxypyrimidin-2-yl)oxy)-3-methoxy-3,3-diphenylpropanoic acid 99+%
(S)-2-((4,6-Dimethoxypyrimidin-2-yl)oxy)-3-methoxy-3,3-diphenylpropanoic acid

Precaución: for Research Use Only

Formula: C₂₂H₂₂N₂O₆
MW: 410.4199 g/mol
MDL Number: MFCD00949063
CAS Number: 171714-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR