tert-Butyl 4-(prop-2-yn-1-yl)piperidine-1-carboxylate 97%

Supplier: Ambeed
Warning

A857125-250MG A857125-1G A857125-5G A857125-100MG
76654-450EA 89.12 USD
76654-450 77569-590 77599-184 77618-410
tert-Butyl 4-(prop-2-yn-1-yl)piperidine-1-carboxylate 97%
tert-Butyl 4-(prop-2-yn-1-yl)piperidine-1-carboxylate

Precaución: For research use only.

Formula: C₁₃H₂₁NO₂
MW: 223.31 g/mol
MDL Number: MFCD17676502
CAS Number: 301185-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR