Methyl 2-amino-3-hydroxy-2-methylpropanoate hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: H-DL-2-Me-Ser-OMe·HCl

A856751-1G A856751-250MG
76654-446EA 157.02 USD
76654-446 76654-448
Methyl 2-amino-3-hydroxy-2-methylpropanoate hydrochloride 97%
Methyl 2-amino-3-hydroxy-2-methylpropanoate hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₂ClNO₃
MW: 169.6067 g/mol
MDL Number: MFCD28400767
CAS Number: 89500-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR